Assay Details
| Gene Symbol: HOXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751450.2 | XP_002751496.1 | 1 - 2 | 1148 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXA10| Gene Symbol: HOXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751450.2 | XP_002751496.1 | 1 - 2 | 1148 | 54 | |
Set Membership: |
|