Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762029.4 | XP_002762075.1 | 1 - 2 | 198 | 79 | |
| XM_017968074.1 | XP_017823563.1 | 1 - 2 | 202 | 79 | ||
More Information
Set Membership: |
|

HCST
Genomic Map