Assay Details
| Gene Symbol: KIF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988815.1 | XP_008987063.1 | 1 - 2 | 865 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF4B| Gene Symbol: KIF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988815.1 | XP_008987063.1 | 1 - 2 | 865 | 82 | |
Set Membership: |
|