Assay Details
| Gene Symbol: TMBIM7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975743.1 | XP_017831232.1 | 6 - 7 | 865 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMBIM7P| Gene Symbol: TMBIM7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975743.1 | XP_017831232.1 | 6 - 7 | 865 | 79 | |