Assay Details
| Gene Symbol: C9H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976590.1 | XP_017832079.1 | 4 - 5 | 241 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9H12orf54| Gene Symbol: C9H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976590.1 | XP_017832079.1 | 4 - 5 | 241 | 75 | |