Assay Details
| Gene Symbol: CORO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008560.2 | XP_009006808.1 | 6 - 7 | 993 | 83 | |
| XM_009008561.2 | XP_009006809.1 | 6 - 7 | 834 | 83 | ||
| XM_017978313.1 | XP_017833802.1 | 5 - 6 | 828 | 83 | ||
| XM_017978314.1 | XP_017833803.1 | 6 - 7 | 1021 | 83 | ||
More Information
Set Membership: |
|

CORO1B
Genomic Map