Assay Details
| Gene Symbol: POU6F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975133.1 | XP_017830622.1 | 1 - 2 | 430 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU6F2| Gene Symbol: POU6F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975133.1 | XP_017830622.1 | 1 - 2 | 430 | 55 | |
Set Membership: |
|