Assay Details
| Gene Symbol: PROCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997185.2 | XP_008995433.1 | 3 - 4 | 544 | 76 | |
| XM_008997189.2 | XP_008995437.1 | 2 - 3 | 297 | 76 | ||
More Information
Set Membership: |
|

PROCA1
Genomic Map