Assay Details
| Gene Symbol: C8G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005723.2 | XP_009003971.1 | 1 - 2 | 416 | 60 | |
| XM_009005726.2 | XP_009003974.1 | 1 - 2 | 433 | 60 | ||
More Information
Set Membership: |
|

C8G
Genomic Map