Assay Details
| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009021.2 | XP_009007269.2 | 4 - 5 | 1542 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF213| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009021.2 | XP_009007269.2 | 4 - 5 | 1542 | 68 | |
Set Membership: |
|