Assay Details
| Gene Symbol: PYCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997914.2 | XP_008996162.1 | 3 - 4 | 303 | 76 | |
| XM_008997915.2 | XP_008996163.1 | 3 - 4 | 285 | 76 | ||
| XM_008997916.2 | XP_008996164.1 | 3 - 4 | 250 | 76 | ||
| XM_008997918.2 | XP_008996166.1 | 3 - 4 | 246 | 76 | ||
| XM_017972933.1 | XP_017828422.1 | 3 - 4 | 285 | 76 | ||
| XM_017972934.1 | XP_017828423.1 | 2 - 3 | 110 | 76 | ||
More Information
Set Membership: |
|

PYCR1
Genomic Map