Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976031.1 | XP_017831520.1 | 3 - 4 | 2046 | 96 | |
| XM_017976032.1 | XP_017831521.1 | 2 - 3 | 984 | 96 | ||
More Information
Set Membership: |
|

DNAJC22
Genomic Map