Assay Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751674.3 | XP_002751720.1 | 2 - 3 | 983 | 101 | |
| XM_009002596.2 | XP_009000844.1 | 3 - 4 | 1238 | 101 | ||
| XM_017975425.1 | XP_017830914.1 | 2 - 3 | 1216 | 101 | ||
More Information
Set Membership: |
|

LRRC17
Genomic Map