Assay Details
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009002915.2 | XP_009001163.1 | 3 - 4 | 2386 | 81 | |
XM_009002918.2 | XP_009001166.1 | 2 - 3 | 519 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
