Assay Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743349.3 | XP_002743395.1 | 4 - 5 | 763 | 58 | |
| XM_009003517.1 | XP_009001765.1 | 4 - 5 | 453 | 58 | ||
| XM_017976526.1 | XP_017832015.1 | 4 - 5 | 402 | 58 | ||
More Information
Set Membership: |
|

COQ4
Genomic Map