Assay Details
| Gene Symbol: NEIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745196.3 | XP_002745242.1 | 2 - 3 | 400 | 74 | |
| XM_008992427.2 | XP_008990675.1 | 2 - 3 | 400 | 74 | ||
More Information
Set Membership: |
|

NEIL3
Genomic Map