Assay Details
Gene Symbol: CCDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762259.3 | XP_002762305.1 | 1 | 2117 | 88 | |
XM_008988268.2 | XP_008986516.1 | 2 | 2069 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
