Assay Details
| Gene Symbol: FGF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755643.3 | XP_002755689.1 | 1 - 2 | 660 | 101 | |
| XM_017978353.1 | XP_017833842.1 | 1 - 2 | 341 | 101 | ||
More Information
Set Membership: |
|

FGF4
Genomic Map