Assay Details
| Gene Symbol: KCNK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807466.3 | XP_002807512.3 | 2 - 3 | 351 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK18| Gene Symbol: KCNK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807466.3 | XP_002807512.3 | 2 - 3 | 351 | 91 | |
Set Membership: |
|