Assay Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988771.2 | XP_008987019.1 | 3 - 4 | 301 | 84 | |
| XM_008988772.2 | XP_008987020.1 | 2 - 3 | 316 | 84 | ||
| XM_008988773.2 | XP_008987021.1 | 3 - 4 | 449 | 84 | ||
| XM_017967699.1 | XP_017823188.1 | 2 - 3 | 243 | 84 | ||
More Information
Set Membership: |
|

ZNF550
Genomic Map