Assay Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988771.2 | XP_008987019.1 | 4 - 5 | 398 | 64 | |
| XM_008988772.2 | XP_008987020.1 | 3 - 4 | 413 | 64 | ||
| XM_008988773.2 | XP_008987021.1 | 4 - 5 | 546 | 64 | ||
| XM_017967699.1 | XP_017823188.1 | 3 - 4 | 340 | 64 | ||
More Information
Set Membership: |
|

ZNF550
Genomic Map