Assay Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988771.2 | XP_008987019.1 | 5 - 6 | 1922 | 80 | |
| XM_008988772.2 | XP_008987020.1 | 4 - 5 | 1937 | 80 | ||
| XM_008988773.2 | XP_008987021.1 | 5 - 6 | 2070 | 80 | ||
| XM_017967699.1 | XP_017823188.1 | 4 - 5 | 1864 | 80 | ||
More Information
Set Membership: |
|

ZNF550
Genomic Map