Assay Details
| Gene Symbol: APOBEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003530.1 | XP_009001778.1 | 3 - 4 | 505 | 89 | |
| XM_017975929.1 | XP_017831418.1 | 2 - 3 | 473 | 89 | ||
More Information
Set Membership: |
|

APOBEC1
Genomic Map