Assay Details
| Gene Symbol: GNRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995653.2 | XP_008993901.1 | 3 - 4 | 2666 | 92 | |
| XM_008995654.2 | XP_008993902.1 | 3 - 4 | 2663 | 92 | ||
| XM_017971911.1 | XP_017827400.1 | 3 - 4 | 1217 | 92 | ||
More Information
Set Membership: |
|

GNRH2
Genomic Map