Assay Details
| Gene Symbol: SCFD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745846.3 | XP_002745892.1 | 4 - 5 | 1478 | 72 | |
| XM_008993346.1 | XP_008991594.1 | 4 - 5 | 1478 | 72 | ||
| XM_017970562.1 | XP_017826051.1 | 4 - 5 | 1478 | 72 | ||
More Information
Set Membership: |
|

SCFD2
Genomic Map