Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983430.2 | XP_008981678.1 | 1 - 2 | 423 | 92 | |
| XM_008983431.2 | XP_008981679.1 | 1 - 2 | 423 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH5| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983430.2 | XP_008981678.1 | 1 - 2 | 423 | 92 | |
| XM_008983431.2 | XP_008981679.1 | 1 - 2 | 423 | 92 | ||