Assay Details
| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753676.3 | XP_002753722.1 | 2 - 3 | 515 | 62 | |
| XM_009005832.1 | XP_009004080.1 | 2 - 3 | 405 | 62 | ||
| XM_017977030.1 | XP_017832519.1 | 2 - 3 | 515 | 62 | ||
More Information
Set Membership: |
|

DHRS1
Genomic Map