Assay Details
| Gene Symbol: LRRC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971567.1 | XP_017827056.1 | 8 - 9 | 1702 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC63| Gene Symbol: LRRC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971567.1 | XP_017827056.1 | 8 - 9 | 1702 | 65 | |
Set Membership: |
|