Assay Details
| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756927.3 | XP_002756973.2 | 2 - 3 | 969 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PURG| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756927.3 | XP_002756973.2 | 2 - 3 | 969 | 112 | |
Set Membership: |
|