Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732800.3 | XP_003732848.2 | 3 - 4 | 1083 | 93 | |
| XM_008995630.2 | XP_008993878.1 | 3 - 4 | 1083 | 93 | ||
More Information
Set Membership: |
|

SIRPB2
Genomic Map