Assay Details
| Gene Symbol: ALPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963608.1 | XP_017819097.1 | 7 - 8 | 6006 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALPK2| Gene Symbol: ALPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963608.1 | XP_017819097.1 | 7 - 8 | 6006 | 63 | |
Set Membership: |
|