Assay Details
| Gene Symbol: LOC100396521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971577.1 | XP_017827066.1 | 1 - 2 | 120 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100396521| Gene Symbol: LOC100396521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971577.1 | XP_017827066.1 | 1 - 2 | 120 | 80 | |
Set Membership: |
|