Assay Details
Gene Symbol: TVP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017978769.1 | XP_017834258.1 | 4 - 5 | 864 | 62 | |
XM_017978770.1 | XP_017834259.1 | 4 - 5 | 864 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
