Assay Details
| Gene Symbol: MRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743484.4 | XP_002743530.2 | 3 - 4 | 430 | 99 | |
| XM_009005214.2 | XP_009003462.1 | 2 - 3 | 241 | 99 | ||
More Information
Set Membership: |
|

MRPS2
Genomic Map