Assay Details
| Gene Symbol: C9H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752699.3 | XP_002752745.2 | 3 - 4 | 700 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9H12orf56| Gene Symbol: C9H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752699.3 | XP_002752745.2 | 3 - 4 | 700 | 77 | |
Set Membership: |
|