Assay Details
| Gene Symbol: LOC100414085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757475.2 | XP_002757521.1 | 6 - 7 | 830 | 79 | |
| XM_017964121.1 | XP_017819610.1 | 6 - 7 | 834 | 79 | ||
More Information
Set Membership: |
|

LOC100414085
Genomic Map