Assay Details
| Gene Symbol: UBE2U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001395.2 | XP_008999643.1 | 1 - 2 | 512 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2U| Gene Symbol: UBE2U | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001395.2 | XP_008999643.1 | 1 - 2 | 512 | 87 | |
Set Membership: |
|