Assay Details
| Gene Symbol: KRTAP11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761355.1 | XP_002761401.1 | 1 | 313 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP11-1| Gene Symbol: KRTAP11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761355.1 | XP_002761401.1 | 1 | 313 | 90 | |
Set Membership: |
|