Assay Details
| Gene Symbol: GPR55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973790.1 | XP_017829279.1 | 1 | 1823 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR55| Gene Symbol: GPR55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973790.1 | XP_017829279.1 | 1 | 1823 | 67 | |
Set Membership: |
|