Assay Details
| Gene Symbol: CELA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750314.2 | XP_002750360.1 | 7 - 8 | 810 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CELA2A| Gene Symbol: CELA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750314.2 | XP_002750360.1 | 7 - 8 | 810 | 69 | |
Set Membership: |
|