Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758116.4 | XP_002758162.1 | 2 - 3 | 228 | 67 | |
| XM_002758117.3 | XP_002758163.1 | 2 - 3 | 228 | 67 | ||
More Information
Set Membership: |
|

AP2M1
Genomic Map