Assay Details
| Gene Symbol: IL9R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763465.3 | XP_002763511.1 | 7 - 8 | 1063 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL9R| Gene Symbol: IL9R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763465.3 | XP_002763511.1 | 7 - 8 | 1063 | 63 | |
Set Membership: |
|