Assay Details
| Gene Symbol: SERPINB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979932.2 | XP_008978180.1 | 4 - 5 | 566 | 69 | |
| XM_017963625.1 | XP_017819114.1 | 4 - 5 | 506 | 69 | ||
More Information
Set Membership: |
|

SERPINB12
Genomic Map