Assay Details
| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992652.2 | XP_008990900.1 | 7 - 8 | 643 | 77 | |
| XM_008992653.2 | XP_008990901.1 | 6 - 7 | 535 | 77 | ||
| XM_017970222.1 | XP_017825711.1 | 7 - 8 | 498 | 77 | ||
More Information
Set Membership: |
|

GYPA
Genomic Map