Assay Details
| Gene Symbol: FAM19A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964247.1 | XP_017819736.1 | 1 - 2 | 255 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM19A5| Gene Symbol: FAM19A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964247.1 | XP_017819736.1 | 1 - 2 | 255 | 69 | |
Set Membership: |
|