Assay Details
| Gene Symbol: TMEM247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757780.3 | XP_002757826.1 | 2 - 3 | 2130 | 116 | |
| GenBank mRNA | HX662972.1 | - | 2 - 3 | 515 | 116 | |
More Information
Set Membership: |
|

TMEM247
Genomic Map