Assay Details
| Gene Symbol: FCRLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984793.2 | XP_008983041.1 | 4 - 5 | 575 | 65 | |
| XM_008984794.2 | XP_008983042.1 | 3 - 4 | 789 | 65 | ||
| XM_008984795.2 | XP_008983043.1 | 3 - 4 | 579 | 65 | ||
More Information
Set Membership: |
|

FCRLA
Genomic Map