Assay Details
| Gene Symbol: STPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970409.1 | XP_017825898.1 | 2 - 3 | 560 | 70 | |
| XM_017970410.1 | XP_017825899.1 | 2 - 3 | 560 | 70 | ||
More Information
Set Membership: |
|

STPG2
Genomic Map