Assay Details
| Gene Symbol: EGLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988050.2 | XP_008986298.1 | 1 | 1282 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EGLN2| Gene Symbol: EGLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988050.2 | XP_008986298.1 | 1 | 1282 | 126 | |
Set Membership: |
|