Assay Details
| Gene Symbol: LOC100389624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746231.4 | XP_002746277.1 | 1 - 2 | 3205 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100389624| Gene Symbol: LOC100389624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746231.4 | XP_002746277.1 | 1 - 2 | 3205 | 127 | |
Set Membership: |
|